Learn More
Invitrogen™ Rabbit anti-Rat IgG (H+L) Secondary Antibody
Rabbit Polyclonal Secondary Antibody
Brand: Invitrogen™ 31218
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31218 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31218 reacts with the heavy chains of rat IgG and with the light chains common to most rat immunoglobulins, but does not react against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Country of Origin: USA.
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Rat IgG (H+L) | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| 2 mg | |
| Secondary | |
| 4°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 2.4 mg/mL | |
| PBS with no preservative; pH 7.6 | |
| Antigen affinity chromatography | |
| RUO | |
| Rat | |
| Secondary Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.