Learn More
RANBP17 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant RANBP17.
Marke: Abnova PAB22304.100uL
Beschreibung
The transport of protein and large RNAs through the nuclear pore complexes (NPC) is an energy-dependent and regulated process. The import of proteins with a nuclear localization signal (NLS) is accomplished by recognition of one or more clusters of basic amino acids by the importin-alpha/beta complex; see MIM 600685 and MIM 602738. The small GTPase RAN (MIM 601179) plays a key role in NLS-dependent protein import. RAN-binding protein-17 is a member of the importin-beta superfamily of nuclear transport receptors.[supplied by OMIM
Sequence: MDGELSCRVFQLISLMDTGLPRCCNEKIELAILWFLDQFRKTYVGDQLQRTSKVYARMSEVLGITDDNHVLETFMTSpezifikation
| RANBP17 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant RANBP17. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| FLJ32916 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 64901 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user. | |
| RANBP17 | |
| RANBP17 | |
| Recombinant protein corresponding to amino acids of human RANBP17. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only