Learn More
RAD9A (phospho S272), Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of RAD9A.
Marca: Abnova PAB0581.400uL
Descrizione
This gene product is highly similar to Schizosaccharomyces pombe rad9, a cell cycle checkpoint protein required for cell cycle arrest and DNA damage repair in response to DNA damage. This protein is found to possess 3' to 5' exonuclease activity, which may contribute to its role in sensing and repairing DNA damage. It forms a checkpoint protein complex with RAD1 and HUS1. This complex is recruited by checkpoint protein RAD17 to the sites of DNA damage, which is thought to be important for triggering the checkpoint-signaling cascade. Use of alternative polyA sites has been noted for this gene. [provided by RefSeq]
Specifica
| RAD9A | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of RAD9A. | |
| In PBS (0.09% sodium azide) | |
| RAD9 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 5883 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:10-100) The optimal working dilution should be determined by the end user. | |
| RAD9A | |
| RAD9A | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S272 of human RAD9A. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
For Research Use Only