missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD9 (phospho S1260), Rabbit anti-Yeast, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of RAD9.
Brand: Abnova PAB11310.100ug
This item is not returnable.
View return policy
Specifications
| RAD9 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of RAD9. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| RAD9 | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S1260 of Saccharomyces cerevisiae RAD9. | |
| RUO | |
| 851803 | |
| Yeast | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| Sandwich ELISA (1:5000) Western Blot (1:100-1:500) The optimal working dilution should be determined by the end user. | |
| RAD9 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This phospho specific polyclonal antibody reacts with phosphorylated pS1260 of yeast Rad9. Reactivity with non-phosphorylated yeast Rad9 is minimal by ELISA and immunoblotting. No reactivity is expected against the human or mouse analogs of RAD9. Cross reactivity may occur with auto-phosphorylated Rad53 kinase. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction