Learn More
RAD52 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of RAD52.
Brand: Abnova PAB10230.100ug
Description
The protein encoded by this gene shares similarity with Saccharomyces cerevisiae Rad52, a protein important for DNA double-strand break repair and homologous recombination. This gene product was shown to bind single-stranded DNA ends, and mediate the DNA-DNA interaction necessary for the annealing of complementary DNA strands. It was also found to interact with DNA recombination protein RAD51, which suggested its role in RAD51 related DNA recombination and repair. [provided by RefSeq
Specifications
| RAD52 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of RAD52. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| RAD52 | |
| A synthetic peptide corresponding to amino acids 360-375 of human RAD52. | |
| RUO | |
| 5893 | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:5000-1:20000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
| RAD52 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Reactivity occurs against human RAD52 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only