Learn More
RAD51L1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of RAD51L1.
Brand: Abnova PAB26924.100uL
Description
The protein encoded by this gene is a member of the RAD51 protein family. RAD51 family members are evolutionarily conserved proteins essential for DNA repair by homologous recombination. This protein has been shown to form a stable heterodimer with the family member RAD51C, which further interacts with the other family members, such as RAD51, XRCC2, and XRCC3. Overexpression of this gene was found to cause cell cycle G1 delay and cell apoptosis, which suggested a role of this protein in sensing DNA damage. At least three alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq
Specifications
| RAD51L1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| RAD51L1 | |
| RAD51L1 | |
| A synthetic peptide corresponding to human RAD51L1. | |
| 100 μL | |
| Primary | |
| RAD51L1 polyclonal antibody detects endogenous levels of RAD51L1 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of RAD51L1. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| MGC34245/R51H2/RAD51B/REC2/hREC2 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 5890 | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only