Learn More
RAD21 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant RAD21.
Brand: Abnova PAB21266.100uL
Description
The protein encoded by this gene is highly similar to the gene product of Schizosaccharomyces pombe rad21, a gene involved in the repair of DNA double-strand breaks, as well as in chromatid cohesion during mitosis. This protein is a nuclear phospho-protein, which becomes hyperphosphorylated in cell cycle M phase. The highly regulated association of this protein with mitotic chromatin specifically at the centromere region suggests its role in sister chromatid cohesion in mitotic cells. [provided by RefSeq
Sequence: KLMMWKETGGVEKLFSLPAQPLWNNRLLKLFTRCLTPLVPEDLRKRRKGGEADNLDEFLKEFENPEVPREDQQQQHQQRDVIDEPIIEEPSRLQESVMEASRTNIDESAMPPPPPQGVKRKAGQIDPEPVMPPQQVEQMEIPPVELSpecifications
| RAD21 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant RAD21. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| FLJ25655/FLJ40596/HR21/HRAD21/KIAA0078/MCD1/NXP1/SCC1/hHR21 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 5885 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:500-1:1000) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| RAD21 | |
| RAD21 | |
| Recombinant protein corresponding to amino acids of human RAD21. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only