Learn More
RAB3GAP2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant RAB3GAP2.
Brand: Abnova PAB21889.100uL
Description
Members of the RAB3 protein family (see RAB3A; MIM 179490) are implicated in Ca(2+)-dependent exocytosis. RAB3GAP, which is involved in regulation of RAB3 activity, is a heterodimeric complex consisting a 130kDa catalytic subunit (RAB3GAP1; MIM 602536) and a 150kDa noncatalytic subunit (RAB3GAP2) (Nagano et al., 1998 [PubMed 9733780]).[supplied by OMIM
Sequence: EHHSILCSILYAVMRFSLKTVKPLSLFDSKGKNAFFKDLTSIQLLPSGEMDPNFISVRQQFLLKVVSAAVQAQHSATKVKDPTSpecifications
| RAB3GAP2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant RAB3GAP2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| DKFZp434D245/FLJ14579/KIAA0839/RAB3-GAP150/RAB3GAP150/p150 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 25782 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:10-1:20) The optimal working dilution should be determined by the end user. | |
| RAB3GAP2 | |
| RAB3GAP2 | |
| Recombinant protein corresponding to amino acids of human RAB3GAP2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only