Learn More
RAB11FIP3 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant RAB11FIP3.
Brand: Abnova PAB22750.100uL
Description
Proteins of the large Rab GTPase family (see RAB1A; MIM 179508) have regulatory roles in the formation, targeting, and fusion of intracellular transport vesicles. RAB11FIP3 is one of many proteins that interact with and regulate Rab GTPases (Hales et al., 2001 [PubMed 11495908]).[supplied by OMIM
Sequence: QVKDLTKYLDPSGLGVISFEDFYQGITAIRNGDPDGQCYGGVASAQDEEPLACPDEFDDFVTYEANEVTDSAYMGSESTYSECETFSpecifications
| RAB11FIP3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant RAB11FIP3. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| KIAA0665/Rab11-FIP3 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 9727 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:500-1:1000) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| RAB11FIP3 | |
| RAB11FIP3 | |
| Recombinant protein corresponding to amino acids of human RAB11FIP3. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only