Learn More
QRFP, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant QRFP.
Brand: Abnova PAB28115.100uL
Description
The P518 precursor protein can be processed into several RF (arg-phe)-amide peptides, including P518. RF-amide peptides share a common C-terminal motif and are involved in cell signaling through G protein-coupled receptors (Jiang et al., 2003 [PubMed 12714592]).[supplied by OMIM
Sequence: GREHAGCRFRFGRQDEGSEATGFLPAAGEKTSGPLGNLAEELNGYSRKKGGFSFRSpecifications
| QRFP | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant QRFP. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| 26RFa/MGC119794/P518 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 347148 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| QRFP | |
| QRFP | |
| Recombinant protein corresponding to amino acids of recombinant QRFP. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only