Learn More
PYGM Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PYGM.
Brand: Abnova PAB16213.50ug
Description
This gene encodes a muscle enzyme involved in glycogenolysis. Highly similar enzymes encoded by different genes are found in liver and brain. Mutations in this gene are associated with McArdle disease (myophosphorylase deficiency), a glycogen storage disease of muscle. Alternative splicing results in multiple transcript variants
Specifications
| PYGM | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PYGM. | |
| In PBS (0.09% sodium azide) | |
| PYGM | |
| A synthetic peptide (conjugated with KLH) corresponding to human PYGM. | |
| 50 μg | |
| Primary | |
| C-terminal domain of human . | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (20 ug/mL) The optimal working dilution should be determined by the end user. | |
| PYGM | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 5837 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only