missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTTG1/PTTG2/PTTG3P, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PTTG1/PTTG2/PTTG3P.
Brand: Abnova PAB24878.100uL
This item is not returnable.
View return policy
Specifications
| PTTG1/PTTG2/PTTG3P | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| EAP1/HPTTG/MGC126883/MGC138276/PTTG/TUTR1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 9232/10744/26255 | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of PTTG1/PTTG2/PTTG3P. | |
| In 1X PBS, pH 7.2 (0.05% sodium azide) | |
| PTTG1 | |
| A synthetic peptide corresponding to PTTG1/PTTG2/PTTG3P. | |
| 100 μL | |
| Primary | |
| PTTG1/PTTG2/PTTG3P polyclonal antibody detects endogenous levels of PTTG1/PTTG2/PTTG3P protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction