Learn More
PTPRU, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant PTPRU.
Brand: Abnova PAB23587.100uL
Description
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP possesses an extracellular region, a single transmembrane region, and two tandem intracellular catalytic domains, and thus represents a receptor-type PTP. The extracellular region contains a meprin-A5 antigen-PTP (MAM) domain, Ig-like and fibronectin type III-like repeats. This PTP was thought to play roles in cell-cell recognition and adhesion. Studies of the similar gene in mice suggested the role of this PTP in early neural development. The expression of this gene was reported to be regulated by phorbol myristate acetate (PMA) or calcium ionophore in Jurkat T lymphoma cells. Three alternatively spliced transcript variants, which encode distinct proteins, have been reported. [provided by RefSeq
Sequence: TFEEASDPAVPCEYSQAQYDDFQWEQVRIHPGTRAPADLPHGSYLMVNTSQHAPGQRAHVIFQSLSENDTHCVQFSYFLYSRDGSpecifications
| PTPRU | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant PTPRU. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| FLJ37530/FMI/GLEPP1/PCP-2/PTP/PTP-J/PTP-PI/PTPPSI/PTPRO/PTPU2/R-PTP-PSI/hPTP-J | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 10076 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| PTPRU | |
| PTPRU | |
| Recombinant protein corresponding to amino acids of human PTPRU. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only