Learn More
PTPRC, Rabbit, Clone: E19-G, Abnova™
Rabbit monoclonal antibody raised against synthetic peptide of PTPRC.
Brand: Abnova MAB1649.100uL
Description
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP contains an extracellular domain, a single transmembrane segment and two tandem intracytoplasmic catalytic domains, and thus belongs to receptor type PTP. This gene is specifically expressed in hematopoietic cells. This PTP has been shown to be an essential regulator of T- and B-cell antigen receptor signaling. It functions through either direct interaction with components of the antigen receptor complexes, or by activating various Src family kinases required for the antigen receptor signaling. This PTP also suppresses JAK kinases, and thus functions as a regulator of cytokine receptor signaling. Four alternatively spliced transcripts variants of this gene, which encode distinct isoforms, have been reported. [provided by RefSeq
Specifications
| PTPRC | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:1000) The optimal working dilution should be determined by the end user. | |
| PTPRC | |
| PTPRC | |
| A synthetic peptide corresponding to C-terminus of human PTPRC. | |
| RUO | |
| 5788 | |
| Store at 4°C. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| E19-G | |
| Rabbit monoclonal antibody raised against synthetic peptide of PTPRC. | |
| In 20mM Tris-HCl, pH 8.0 (20mg/mL BSA, 0.05% sodium azide) | |
| B220/CD45/CD45R/GP180/LCA/LY5/T200 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only