Learn More
PTPN11 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PTPN11.
Brand: Abnova PAB12287.100ug
Description
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP contains two tandem Src homology-2 domains, which function as phospho-tyrosine binding domains and mediate the interaction of this PTP with its substrates. This PTP is widely expressed in most tissues and plays a regulatory role in various cell signaling events that are important for a diversity of cell functions, such as mitogenic activation, metabolic control, transcription regulation, and cell migration. Mutations in this gene are a cause of Noonan syndrome as well as acute myeloid leukemia. [provided by RefSeq
Specifications
| PTPN11 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PTPN11. | |
| Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
| BC008692 | |
| PTPN11 | |
| A synthetic peptide corresponding to amino acids at C-terminus of human PTPN11. | |
| 100 μg | |
| Primary | |
| Identical to the related rat sequence. | |
| Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water to a concentration of 500μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| PTPN11 | |
| BPTP3/CFC/MGC14433/NS1/PTP-1D/PTP2C/SH-PTP2/SH-PTP3/SHP2 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 5781 | |
| Human, Mouse, Rat | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only