Learn More
PTGFR Rabbit anti-Horse, Human, Monkey, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PTGFR.
Brand: Abnova PAB26438.50ug
Description
The protein encoded by this gene is member of the G-protein coupled receptor family. This protein is a receptor for prostaglandin F2-alpha (PGF2-alpha), which is known to be a potent luteolytic agent, and may also be involved in modulating intraocular pressure and smooth muscle contraction in uterus. Knockout studies in mice suggest that the interaction of PGF2-alpha with this receptor may initiate parturition in ovarian luteal cells and thus induce luteolysis. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
| PTGFR | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PTGFR. | |
| In PBS (0.09% sodium azide) | |
| FP/MGC120498/MGC46203 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 5737 | |
| Equine, Human, Monkey | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-8 ug/mL) The optimal working dilution should be determined by the end user. | |
| PTGFR | |
| PTGFR | |
| A synthetic peptide corresponding to 16 amino acids at 2nd extracellular domain of human PTGFR. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only