Learn More
PTAFR, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant PTAFR.
Brand: Abnova PAB22031.100uL
Description
PTAFR shows structural characteristics of the rhodopsin (MIM 180380) gene family and binds platelet-activating factor (PAF). PAF is a phospholipid (1-0-alkyl-2-acetyl-sn-glycero-3-phosphorylcholine) that has been implicated as a mediator in diverse pathologic processes, such as allergy, asthma, septic shock, arterial thrombosis, and inflammatory processes.[supplied by OMIM
Sequence: LDSTNTVPDSAGSGNVTRCFEHYEKGSSpecifications
| PTAFR | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant PTAFR. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| PAFR | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 5724 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| PTAFR | |
| PTAFR | |
| Recombinant protein corresponding to amino acids of human PTAFR. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only