missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMD14, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant PSMD14.
Brand: Abnova PAB28572.100uL
This item is not returnable.
View return policy
Description
PSMD14 is a component of the 26S proteasome, a multiprotein complex that degrades proteins targeted for destruction by the ubiquitin pathway (Spataro et al., 1997 [PubMed 9374539]).[supplied by OMIM
Sequence: AVDTAEQVYISSLALLKMLKHGRAGVPMEVMGLMLGEFVDDYTVRVIDVFAMPQSGTGVSVEAVDPVFQAKMLDMLKQTGRPEMVVGWYHSHPGFGCWLSGVDINTQQSFEALSERAVAVVVDPIQSVKGKVVIDASpecifications
| PSMD14 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant PSMD14. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| PAD1/POH1/rpn11 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 10213 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (Human: 1:100-1:250, Mouse/Rat: 1:100-1:500) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1:500) Immunofluorescence (1-4 ug/ml) The optimal working dilution should be determined by the end user. | |
| PSMD14 | |
| PSMD14 | |
| Recombinant protein corresponding to human PSMD14. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction