missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRMT5 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PRMT5.
Brand: Abnova PAB0221.200uL
This item is not returnable.
View return policy
Specifications
| PRMT5 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PRMT5. | |
| In PBS (0.05% BSA, 0.05% sodium azide) | |
| HRMT1L5/IBP72/JBP1/SKB1/SKB1Hs | |
| Rabbit | |
| 200 μL | |
| Primary | |
| Human | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| PRMT5 | |
| PRMT5 | |
| A mixture of synthetic peptides corresponding to human PRMT5. | |
| RUO | |
| 10419 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction