missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRLHR Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PRLHR.
Brand: Abnova PAB16337.50ug
This item is not returnable.
View return policy
Description
PRLHR is a 7-transmembrane domain receptor for prolactin-releasing hormone (PRLH; MIM 602663) that is highly expressed in anterior pituitary (Ozawa et al., 2002 [PubMed 11923475]).[supplied by OMIM
Specifications
| PRLHR | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PRLHR. | |
| In PBS (0.09% sodium azide) | |
| GPR10/GR3/MGC126539/MGC126541/PrRPR | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 2834 | |
| Human, Mouse, Rat | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (20 ug/mL) The optimal working dilution should be determined by the end user. | |
| PRLHR | |
| PRLHR | |
| A synthetic peptide (conjugated with KLH) corresponding to human PRLHR. | |
| 50 μg | |
| Primary | |
| 2nd extracellular domain of human. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction