missing translation for 'onlineSavingsMsg'
Learn More

PRKDC (phospho T2609), Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic phosphopeptide of PRKDC.

Brand:  Abnova PAB7737.100uL

Product Code. 16224480

  • £349.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The PRKDC gene encodes the catalytic subunit of a nuclear DNA-dependent serine/threonine protein kinase (DNA-PK). The second component is the autoimmune antigen Ku (MIM 152690), which is encoded by the G22P1 gene on chromosome 22q. On its own, the catalytic subunit of DNA-PK is inactive and relies on the G22P1 component to direct it to the DNA and trigger its kinase activity; PRKDC must be bound to DNA to express its catalytic properties.[supplied by OMIM

Specifications

Specifications

PRKDC
Polyclonal
Unconjugated
Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
PRKDC
PRKDC
Synthetic phosphopeptide corresponding to residues surrounding T2609 of human PRKDC.
100 μL
Primary
Human
Liquid
Western Blot
1 mg/mL
Rabbit polyclonal antibody raised against synthetic phosphopeptide of PRKDC.
In PBS (without Mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
DNA-PKcs/DNAPK/DNPK1/HYRC/HYRC1/XRCC7/p350
Rabbit
Immunoaffinity purification
RUO
5591
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only