missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRKACA/PRKACB, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human PRKACA/PRKACB.
Brand: Abnova PAB29264.100uL
This item is not returnable.
View return policy
Specifications
| PRKACA/PRKACB | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of human PRKACA/PRKACB. | |
| In PBS (without Mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| PRKACA | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding T197 of human PRKACA/PRKACB. | |
| 100 μL | |
| Primary | |
| PRKACA/PRKACB (phospho T197) polyclonal antibody detects endogenous levels of human PRKACA/PRKACB only when phosphorylated at threonine 197. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunofluorescence (1:100∼1:200) Immunohistochemistry (1:50∼1:100) Western Blot (1:500∼1:1000) The optimal working dilution should be determined by the end user. | |
| MGC102831/MGC48865/PKACA | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 5566/5567 | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction