Learn More
PRKAB1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PRKAB1.
Brand: Abnova PAB19840.100ug
Description
The protein encoded by this gene is a regulatory subunit of the AMP-activated protein kinase (AMPK). AMPK is a heterotrimer consisting of an alpha catalytic subunit, and noncatalytic beta and gamma subunits. AMPK is an important energy-sensing enzyme that monitors cellular energy status. In response to cellular metabolic stresses, AMPK is activated, and thus phosphorylates and inactivates acetyl-CoA carboxylase (ACC) and beta-hydroxy beta-methylglutaryl-CoA reductase (HMGCR), key enzymes involved in regulating de novo biosynthesis of fatty acid and cholesterol. This subunit may be a positive regulator of AMPK activity. The myristoylation and phosphorylation of this subunit have been shown to affect the enzyme activity and cellular localization of AMPK. This subunit may also serve as an adaptor molecule mediating the association of the AMPK complex. [provided by RefSeq
Specifications
| PRKAB1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PRKAB1. | |
| In serum (0.02% sodium azide) | |
| AMPK/HAMPKb/MGC17785 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:40000-1:80000) Western Blot (1:200-1000) The optimal working dilution should be determined by the end user. | |
| PRKAB1 | |
| PRKAB1 | |
| A synthetic peptide corresponding to 15 amino acids near N-terminus of human PRKAB1. | |
| RUO | |
| 5564 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only