Learn More
PRKAA1 (phospho T172), Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of PRKAA1.
Brand: Abnova PAB12602.100ug
Description
The protein encoded by this gene belongs to the ser/thr protein kinase family. It is the catalytic subunit of the 5'-prime-AMP-activated protein kinase (AMPK). AMPK is a cellular energy sensor conserved in all eukaryotic cells. The kinase activity of AMPK is activated by the stimuli that increase the cellular AMP/ATP ratio. AMPK regulates the activities of a number of key metabolic enzymes through phosphorylation. It protects cells from stresses that cause ATP depletion by switching off ATP-consuming biosynthetic pathways. Alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq]
Sequence: LRTSSpecifications
| PRKAA1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of PRKAA1. | |
| In TBS, pH 7.2 (BSA, 10% Proclin300) | |
| AMPK/AMPKa1/MGC33776/MGC57364 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody recognizes PRKAA1 with a phosphorylated site of Thr 172. It does not cross-react with non-phosphospecific peptide. | |
| Store at 4°C. For long term storage store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) The optimal working dilution should be determined by the end user. | |
| PRKAA1 | |
| PRKAA1 | |
| Synthetic phosphopeptide corresponding to residues surrounding T172 of human PRKAA1. | |
| RUO | |
| 5562 | |
| Bovine, Canine, Chicken, Human, Insect, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only