missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Prion protein, Clone: V21-V, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP1-71678-0.05ML
This item is not returnable.
View return policy
Description
PrPSc Monoclonal specifically detects PrPSc in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, ELISA, Immunoprecipitation.
Specifications
| Prion protein | |
| Monoclonal | |
| 1.5mg/mL | |
| Western Blot 1:1000, ELISA 1:20000-1:50000, Immunoprecipitation 1:10-1:500 | |
| CD230, CJD, fatal familial insomnia), GSS, prion protein, prion protein (p27-30), prion protein PrP, prion-related protein, PRIPMGC26679 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Bovine |
| Western Blot, ELISA, Immunoprecipitation | |
| V21-V | |
| Unconjugated | |
| 20mM Tris/HCl (pH 8.0) and 10 mg/ml BSA with 0.05% Sodium Azide | |
| PRNP | |
| Peptide derived the middle of the second a-helical domain of human prion protein. | |
| 0.05 mL | |
| Neuroscience, Stem Cell Markers | |
| 5621 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction