Learn More
PRIM1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant PRIM1.
Brand: Abnova PAB23609.100uL
Description
The replication of DNA in eukaryotic cells is carried out by a complex chromosomal replication apparatus, in which DNA polymerase alpha and primase are two key enzymatic components. Primase, which is a heterodimer of a small subunit and a large subunit, synthesizes small RNA primers for the Okazaki fragments made during discontinuous DNA replication. The protein encoded by this gene is the small, 49kDa primase subunit. [provided by RefSeq
Sequence: ELVFDIDMTDYDDVRRCCSSADICPKCWTLMTMAIRIIDRALKEDFGFKHRLWVYSGRRGVHCWVCDESVRKLSSAVRSGSpecifications
| PRIM1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant PRIM1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| MGC12308/p49 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 5557 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| PRIM1 | |
| PRIM1 | |
| Recombinant protein corresponding to amino acids of human PRIM1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only