Learn More
PREX1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant PREX1.
Brand: Abnova PAB20101.100uL
Description
The protein encoded by this gene acts as a guanine nucleotide exchange factor for the RHO family of small GTP-binding proteins (RACs). It has been shown to bind to and activate RAC1 by exchanging bound GDP for free GTP. The encoded protein, which is found mainly in the cytoplasm, is activated by phosphatidylinositol-3,4,5-trisphosphate and the beta-gamma subunits of heterotrimeric G proteins. [provided by RefSeq
Sequence: CAGQCILKVNGSNVMNDGAPEVLEHFQAFRSRREEALGLYQWIYHTHEDAQEARASQEASTEDPSGEQAQEEDQADSAFPLLSLGPRLSLCEDSPMVTLTVDNVHLEHGVVYEYVSTAGVRCHVLEKIVEPSpecifications
| PREX1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant PREX1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| KIAA1415/P-REX1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 57580 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| PREX1 | |
| PREX1 | |
| Recombinant protein corresponding to amino acids of human PREX1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only