Learn More
PRCC Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant PRCC.
Brand: Abnova PAB21207.100uL
Description
In a subset of papillary renal cell carcinomas, a t(X;1)(p11;q21) chromosome translocation has been repeatedly reported and is thought to be the cause of the cancer. As a result of the translocation, the transcription factor TFE3 on the X chromosome becomes fused to this gene on chromosome 1. The fused gene results in the fusion of N-terminal proline-rich region of the protein encoded by this gene to the entire TFE3 protein. This protein has been shown to interact with the mitotic checkpoint protein MAD2B, which suggests that the dominant-negative effect of the fusion protein with TFE3 may lead to a mitotic checkpoint defect. Alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq
Sequence: RNRGREEINFVEIKGDDQLSGAQQWMTKSLTEEKTMKSFSKKKGEQPTGQQRRKHQITYLIHQAKERELELKNTWSENKLSRRQTQAKYSpecifications
| PRCC | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant PRCC. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| MGC17178/MGC4723/RCCP1/TPRC | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 5546 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| PRCC | |
| PRCC | |
| Recombinant protein corresponding to amino acids of human PRCC. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
For Research Use Only