missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PQBP1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant PQBP1.
Brand: Abnova PAB28529.100uL
This item is not returnable.
View return policy
Description
PQBP1 is a nuclear polyglutamine-binding protein that contains a WW domain (Waragai et al., 1999 [PubMed 10332029]).[supplied by OMIM
Sequence: PVALQTRLAKRGILKHLEPEPEEEIIAEDYDDDPVDYEATRLEGLPPSWYKVFDPSCGLPYYWNADTDLVSWLSPHDPNSVVTKSAKKLRSSNADAEEKLDRSHDKSDRGHDKSDRSHEKLDRGHDKSDRGHDKSDRDRERGYDKVDSpecifications
| PQBP1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant PQBP1. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| MRX55/MRXS3/MRXS8/NPW38/RENS1/SHS | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 10084 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Immunofluorescence (1-4 ug/ml) Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| PQBP1 | |
| PQBP1 | |
| Recombinant protein corresponding to amino acids of human PQBP1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction