Learn More
PPP2R3A Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant PPP2R3A.
Brand: Abnova PAB22913.100uL
Description
Protein phosphatase 2 (formerly named type 2A) is one of the four major Ser/Thr phosphatases and is implicated in the negative control of cell growth and division. Protein phosphatase 2 holoenzymes are heterotrimeric proteins composed of a structural subunit A, a catalytic subunit C, and a regulatory subunit B. The regulatory subunit is encoded by a diverse set of genes that have been grouped into the B/PR55, B'/PR61, and B″/PR72 families. These different regulatory subunits confer distinct enzymatic specificities and intracellular localizations to the holozenzyme. The product of this gene belongs to the B″ family. The B″ family has been further divided into subfamilies. The product of this gene belongs to the beta subfamily of regulatory subunit B″. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: EQRDPFAVQKDVENDGPEPSDWDRFAAEEYETLVAEESAQAQFQEGFEDYETDEPASPSEFGNKSNKILSASLPEKCGKLQSVDEESpecifications
| PPP2R3A | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant PPP2R3A. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| PPP2R3/PR130/PR72 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 5523 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| PPP2R3A | |
| PPP2R3A | |
| Recombinant protein corresponding to amino acids of human PPP2R3A. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only