Learn More
PPP2CA Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PPP2CA.
Brand: Abnova PAB12283.100ug
Description
This gene encodes the phosphatase 2A catalytic subunit. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. This gene encodes an alpha isoform of the catalytic subunit. [provided by RefSeq
Specifications
| PPP2CA | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PPP2CA. | |
| Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
| PP2Ac/PP2CA/RP-C | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 5515 | |
| Human, Mouse, Rat | |
| Lyophilized |
| Immunocytochemistry, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) Immunocytochemistry (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| PPP2CA | |
| PPP2CA | |
| A synthetic peptide corresponding to amino acids at N-terminus of human PPP2CA. | |
| 100 μg | |
| Primary | |
| Identical to the related rat and mouse sequence. | |
| Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water to a concentration of 500μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only