Learn More
PPP1R15A, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant PPP1R15A.
Brand: Abnova PAB21284.100uL
Beskrivning
This gene is a member of a group of genes whose transcript levels are increased following stressful growth arrest conditions and treatment with DNA-damaging agents. The induction of this gene by ionizing radiation occurs in certain cell lines regardless of p53 status, and its protein response is correlated with apoptosis following ionizing radiation. [provided by RefSeq
Sequence: QPGEDTEEEEDEDSDTGSAEDEREAETSASTPPASAFLKAWVYRPGEDTEEEEDEDVDSEDKEDDSEAALGEAESDPHPSHPDQRAHFRGWGYRPGKETEEEEAAEDWGEAEPCPSpecifikationer
| PPP1R15A | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant PPP1R15A. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| GADD34 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 23645 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user. | |
| PPP1R15A | |
| PPP1R15A | |
| Recombinant protein corresponding to amino acids of human PPP1R15A. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
For Research Use Only