Learn More
PPM1L Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant PPM1L.
Brand: Abnova PAB21255.100uL
Description
PPM1L, or PP2CE, belongs to the PP2C group of serine/threonine phosphatases, which are distinguished from other phosphatases by their structure, absolute requirement for Mg(2+) or Mn(2+), and insensitivity to okadaic acid. PP2Cs regulate stress-activated protein kinase (SAPK; see MIM 601158) signaling cascades that respond to extracellular stimuli (Jin et al., 2004 [PubMed 15560375]).[supplied by OMIM
Sequence: VLCDKDGNAIPLSHDHKPYQLKERKRIKRAGGFISFNGSWRVQGILAMSRSLGDYPLKNLNVVIPDPDILTFSpecifications
| PPM1L | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant PPM1L. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| MGC132545/MGC132547/PP2C-epsilon/PP2CE/PPM1-LIKE | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 151742 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| PPM1L | |
| PPM1L | |
| Recombinant protein corresponding to amino acids of human PPM1L. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only