Learn More
PPARG Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PPARG.
Brand: Abnova PAB14848.100ug
Description
This gene encodes a member of the peroxisome proliferator-activated receptor (PPAR) subfamily of nuclear receptors. PPARs form heterodimers with retinoid X receptors (RXRs) and these heterodimers regulate transcription of various genes. Three subtypes of PPARs are known: PPAR-alpha, PPAR-delta, and PPAR-gamma. The protein encoded by this gene is PPAR-gamma and is a regulator of adipocyte differentiation. Additionally, PPAR-gamma has been implicated in the pathology of numerous diseases including obesity, diabetes, atherosclerosis and cancer. Alternatively spliced transcript variants that encode different isoforms have been described. [provided by RefSeq
Specifications
| PPARG | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PPARG. | |
| Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
| CIMT1/NR1C3/PPARG1/PPARG2/PPARgamma | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 5468 | |
| Human | |
| Lyophilized |
| Western Blot | |
| Unconjugated | |
| Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| PPARG | |
| PPARG | |
| A synthetic peptide corresponding to amino acids at N-terminus of human PPARG. | |
| 100 μg | |
| Primary | |
| Different from the rat and mouse sequence by twoamino acids. | |
| Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water to a concentration of 500μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only