Learn More
PPARG Rabbit anti-Dog, Guinea pig, Hamster, Human, Macaque, Mink, Mouse, Orangutan, Pig, Rabbit, Rat, Squirrel, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PPARG.
Brand: Abnova PAB11247.100ug
Description
This gene encodes a member of the peroxisome proliferator-activated receptor (PPAR) subfamily of nuclear receptors. PPARs form heterodimers with retinoid X receptors (RXRs) and these heterodimers regulate transcription of various genes. Three subtypes of PPARs are known: PPAR-alpha, PPAR-delta, and PPAR-gamma. The protein encoded by this gene is PPAR-gamma and is a regulator of adipocyte differentiation. Additionally, PPAR-gamma has been implicated in the pathology of numerous diseases including obesity, diabetes, atherosclerosis and cancer. Alternatively spliced transcript variants that encode different isoforms have been described. [provided by RefSeq
Specifications
| PPARG | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PPARG. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| CIMT1/NR1C3/PPARG1/PPARG2/PPARgamma | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This affinity purified antibody is directed against human PPAR gamma protein. Expect reactivity against human (gamma1 and gamma2), mouse (gamma1 and gamma2), rat, squirrel, orangutan, duck (gamma1), macaque (gamma1-gamma7), boar (gamma1a-1d), mink, guinea pig, rabbit (gamma1 and gamma3), dog and hamster sources based on 100% homology for the immunogen sequence. No reactivity is expected against other subtypes of PPAR. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:8000-1:32000) The optimal working dilution should be determined by the end user. | |
| PPARG | |
| PPARG | |
| A synthetic peptide corresponding to amino acids 255 -268 of human PPARG. | |
| RUO | |
| 5468 | |
| Canine, Guinea Pig, Hamster, Human, Macaque, Mink, Mouse, Orangutan, Pig, Rabbit, Rat, Squirrel | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only