Learn More
PPAN-P2RY11 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant PPAN-P2RY11
Brand: Abnova PAB28002.100uL
Description
The PPAN-P2RY11 mRNA is a naturally occurring read-through product, a result of intergenic splicing between adjacent genes, PPAN and P2RY11. The encoded fusion protein comprises of sequence sharing identity with each individual gene product. This transcript is found to be ubiquitously expressed and is upregulated by agents inducing granulocytic differentiation. However, its functional significance in vivo remains unclear. [provided by RefSeq
Sequence: RWEMDRGRGRLCDQKFPKTKDKSQGAQARRGPRGASRDGGSpecifications
| PPAN-P2RY11 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant PPAN-P2RY11 | |
| In PBS, pH 7.2, (40% glycerol, 0.02% sodium azide) | |
| PPAN-P2RY11 | |
| Recombinant protein corresponding to amino acids of human PPAN-P2RY11 | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry, Western Blot, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:200 - 1:500) Western Blot (1:100 - 1:250) Immunoflurorescence (1-4 µg/ml) The optimal working dilution should be determined by the end user. | |
| PPAN-P2RY11 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 692312 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only