missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Porcine IgG F(ab')2 Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-72652
This item is not returnable.
View return policy
Description
IgG F(ab')2 Polyclonal antibody specifically detects IgG F(ab')2 in Porcine samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| IgG F(ab')2 | |
| Polyclonal | |
| Alkaline Phosphatase (AP) | |
| immunoglobulin heavy constant gamma 1 (G1m marker) | |
| Swine IgG F(ab')2 fragment | |
| 1 mg | |
| Secondary | |
| Pig | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, ELISA 1:1000 - 1:3000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| No reaction was observed against Swine IgG F(c). | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction