missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Por, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against native Por.
Brand: Abnova PAB11972.50ug
This item is not returnable.
View return policy
Specifications
| Por | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against native Por. | |
| In PBS (0.09% sodium azide, 50% glycerol) | |
| Por | |
| Native purified Por from rat liver. | |
| RUO | |
| 29441 | |
| Avian, Bovine, Canine, Guinea Pig, Hamster, Human, Monkey, Mouse, Pig, Rat, Sheep | |
| Liquid |
| Immunocytochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunocytochemistry (1:500-1:1000) Immunoprecipitation (1:100) The optimal working dilution should be determined by the end user. | |
| Por | |
| Rabbit | |
| 50 μg | |
| Primary | |
| This antibody detects a 78 KDa protein on SDS-PAGE immunoblots, corresponding to the molecular weight of cytochrome P450 reductase. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction