Learn More
POLDIP3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant POLDIP3.
Brand: Abnova PAB21164.100uL
Description
This gene encodes a protein that interacts with the DNA polymerase delta p50 subunit. This protein is a specific target of S6 kinase 1 and regulates cell growth. Two transcript variants that encode different protein isoforms have been identified. [provided by RefSeq
Sequence: SLDELIRKRGAAAKGRLNARPGVGGVRSRVGIQQGLLSQSTRTATFQQRFDARQKIGLSDARLKLGVKDAREKLLQKDARFRIKGKVQDARSpecifications
| POLDIP3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant POLDIP3. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| KIAA1649/PDIP46/SKAR | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 84271 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) Western Blot (1:250-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| POLDIP3 | |
| POLDIP3 | |
| Recombinant protein corresponding to amino acids of human POLDIP3. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only