Learn More
PLD2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human PLD2.
Brand: Abnova PAB29251.100uL
Description
Phosphatidylcholine (PC)-specific phospholipases D (PLDs; EC 3.1.4.4) catalyze the hydrolysis of PC to produce phosphatidic acid and choline. Activation of PC-specific PLDs occurs as a consequence of agonist stimulation of both tyrosine kinase and G protein-coupled receptors. PC-specific PLDs have been proposed to function in regulated secretion, cytoskeletal reorganization, transcriptional regulation, and cell cycle control.[supplied by OMIM
Specifications
| PLD2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of human PLD2. | |
| In PBS, pH 7.4 (without Mg2+ and Ca2+), (50% glycerol, 0.02% sodium azide) | |
| PLD2 | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y169 of human PLD2. | |
| 100 μL | |
| Primary | |
| This antibody detects endogenous levels of PLD2 only when phosphorylated at tyrosine 169. | |
| Store at -20°C. Avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) Western Blot (1:500-1000) The optimal working dilution should be determined by the end user. | |
| PLD2 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 5338 | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only