Learn More
PLCG1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PLCG1.
Brand: Abnova PAB18210.100ug
Description
The protein encoded by this gene catalyzes the formation of inositol 1,4,5-trisphosphate and diacylglycerol from phosphatidylinositol 4,5-bisphosphate. This reaction uses calcium as a cofactor and plays an important role in the intracellular transduction of receptor-mediated tyrosine kinase activators. For example, when activated by SRC, the encoded protein causes the Ras guanine nucleotide exchange factor RasGRP1 to translocate to the Golgi, where it activates Ras. Also, this protein has been shown to be a major substrate for heparin-binding growth factor 1 (acidic fibroblast growth factor)-activated tyrosine kinase. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
| PLCG1 | |
| Polyclonal | |
| Unconjugated | |
| Immunofluorescence (1:500-1:1000) ELISA (1:5000) The optimal working dilution should be determined by the end user. | |
| PLCG1 | |
| PLCG1 | |
| A synthetic peptide corresponding to human PLCG1. | |
| 100 μg | |
| Primary | |
| This antibody is specific to PLCG1. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunofluorescence | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of PLCG1. | |
| In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| PLC-II/PLC1/PLC148/PLCgamma1 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 5335 | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only