missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plat Rabbit anti-Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant Plat.
Brand: Abnova PAB13664.100ug
This item is not returnable.
View return policy
Specifications
| Plat | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant Plat. | |
| In 50mM sodium phosphate buffer, 100mM NaCl, 1mM EDTA, pH 7.4 | |
| AU020998/AW212668/D8Ertd2e/MGC18508/t-PA/tPA | |
| Rabbit | |
| 100 μg | |
| Primary | |
| These polyclonal antibodies are specific to Mouse tPA. They are available as serum, IgG fraction, or affinity purified as well as labeled with biotin and FITC. | |
| Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:100000) Western Blot (1:10000) The optimal working dilution should be determined by the end user. | |
| Plat | |
| Plat | |
| Recombinant protein corresponding to mouse Plat. | |
| RUO | |
| 18791 | |
| Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction