Learn More
PKM2 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PKM2.
Brand: Abnova PAB2038.400uL
Description
This gene encodes a protein involved in glycolysis. The encoded protein is a pyruvate kinase that catalyzes the transfer of a phosphoryl group from phosphoenolpyruvate to ADP, generating ATP and pyruvate. This protein has been shown to interact with thyroid hormone and may mediate cellular metabolic effects induced by thyroid hormones. This protein has been found to bind Opa protein, a bacterial outer membrane protein involved in gonococcal adherence to and invasion of human cells, suggesting a role of this protein in bacterial pathogenesis. Three alternatively spliced transcript variants encoding two distinct isoforms have been reported. [provided by RefSeq
Sequence: NFAMNVGKARGFFKKSpecifications
| PKM2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PKM2. | |
| In PBS (0.09% sodium azide) | |
| CTHBP/MGC3932/OIP3/PK3/PKM/TCB/THBP1 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 5315 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| PKM2 | |
| PKM2 | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human PKM2. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only