missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIWIL2 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PIWIL2.
Brand: Abnova PAB25748.100ug
This item is not returnable.
View return policy
Description
PIWIL2 belongs to the Argonaute family of proteins, which function in development and maintenance of germline stem cells (Sasaki et al., 2003 [PubMed 12906857]).[supplied by OMIM
Specifications
| PIWIL2 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| PIWIL2 | |
| PIWIL2 | |
| A synthetic peptide corresponding to 17 amino acids at internal region of human PIWIL2. | |
| 100 μg | |
| Primary | |
| At least three isoforms of PIWI-L2 are known to exist; this antibody will only detect the two longest isoforms. PIWI-L2 antibody is predicted to not cross-react with other PIWI protein family members. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of PIWIL2. | |
| In PBS (0.02% sodium azide) | |
| FLJ10351/HILI/MGC133049/PIWIL1L/mili | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 55124 | |
| Human, Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction