Learn More
PIK3CD (phospho Y524), Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of PIK3CD.
Brand: Abnova PAB8136.400uL
Description
Phosphoinositide 3-kinases (PI3Ks) phosphorylate the 3-prime OH position of the inositol ring of inositol lipids. See MIM 602838. The class I PI3Ks display a broad phosphoinositide lipid substrate specificity and include p110-alpha (MIM 171834), p110-beta (MIM 602925), and p110-gamma (MIM 601232). p110-alpha and p110-beta interact with SH2/SH3-domain-containing p85 adaptor proteins (see MIM 171833) and with GTP-bound Ras.[supplied by OMIM
Specifications
| PIK3CD | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of PIK3CD. | |
| In PBS (0.09% sodium azide) | |
| p110D | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 5293 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Dot Blot, ELISA | |
| Unconjugated | |
| ELISA (1:1000) Dot Blot (1:500) The optimal working dilution should be determined by the end user. | |
| PIK3CD | |
| PIK3CD | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y524 of human PIK3CD. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only