missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHF2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PHF2.
Brand: Abnova PAB12583.100ug
This item is not returnable.
View return policy
Specifications
| PHF2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PHF2. | |
| In PBS (0.02% sodium azide) | |
| A synthetic peptide corresponding to internal region of PHF2. | |
| RUO | |
| Reacts with residues 70-82 [KHGPGPTPDVKRVC] of the 120 KDa PHF protein. | |
| Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction