missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFTK1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PFTK1.
Brand: Abnova PAB2949.400uL
This item is not returnable.
View return policy
Description
PFTK1 is a member of the CDC2 (MIM 116940)-related protein kinase family (Yang and Chen, 2001 [PubMed 11313143]).[supplied by OMIM
Specifications
| PFTK1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PFTK1. | |
| In PBS (0.09% sodium azide) | |
| KIAA0834/PFTAIRE1 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 5218 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
| PFTK1 | |
| PFTK1 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human PFTK1. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction