Learn More
PFKP Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant PFKP
Brand: Abnova PAB28016.100uL
Description
The PFKP gene encodes the platelet isoform of phosphofructokinase (PFK) (ATP:D-fructose-6-phosphate-1-phosphotransferase, EC 2.7.1.11). PFK catalyzes the irreversible conversion of fructose-6-phosphate to fructose-1,6-bisphosphate and is a key regulatory enzyme in glycolysis. The PFKP gene, which maps to chromosome 10p, is also expressed in fibroblasts. See also the muscle (PFKM; MIM 610681) and liver (PFKL; MIM 171860) isoforms of phosphofructokinase, which map to chromosomes 12q13 and 21q22, respectively. Vora (1981) [PubMed 6451249] determined that full tetrameric phophofructokinase enzyme expressed in platelets can be composed of subunits P4, P3L, and P2L2.[supplied by OMIM
Sequence: SILGTKRVLPGKYLEEIATQMRTHSINALLIIGGFEAYLGLLELSAAREKHEEFCVPMVMVPATVSNNVPGSSpecifications
| PFKP | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant PFKP | |
| In PBS, pH 7.2, (40% glycerol, 0.02% sodium azide) | |
| FLJ40226/PFK-C/PFKF | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 5214 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50 - 1:200) Western Blot (1:100 - 1:250) The optimal working dilution should be determined by the end user. | |
| PFKP | |
| PFKP | |
| Recombinant protein corresponding to amino acids of human PFKP | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only