Learn More
PFKFB1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PFKFB1.
Brand: Abnova PAB4021.400uL
Description
This gene encodes a member of the family of bifunctional 6-phosphofructo-2-kinase:fructose-2,6-biphosphatase enzymes. The enzyme forms a homodimer that catalyzes both the synthesis and degradation of fructose-2,6-biphosphate using independent catalytic domains. Fructose-2,6-biphosphate is an activator of the glycolysis pathway and an inhibitor of the gluconeogenesis pathway. Consequently, regulating fructose-2,6-biphosphate levels through the activity of this enzyme is thought to regulate glucose homeostasis. [provided by RefSeq
Specifications
| PFKFB1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PFKFB1. | |
| In PBS (0.09% sodium azide) | |
| F6PK/HL2K/MGC116715/MGC116717/PFRX | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 5207 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| ELISA (1:1000) Western Blot (1:100-500) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
| PFKFB1 | |
| PFKFB1 | |
| A synthetic peptide (conjugated with KLH) corresponding to internal region of human PFKFB1. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only