Learn More
PDSS1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant PDSS1.
Brand: Abnova PAB23207.100uL
Description
The protein encoded by this gene is an enzyme that elongates the prenyl side-chain of coenzyme Q, or ubiquinone, one of the key elements in the respiratory chain. The gene product catalyzes the formation of all trans-polyprenyl pyrophosphates from isopentyl diphosphate in the assembly of polyisoprenoid side chains, the first step in coenzyme Q biosynthesis. The protein may be peripherally associated with the inner mitochondrial membrane, though no transit peptide has been definitively identified to date. Defects in this gene are a cause of coenzyme Q10 deficiency. [provided by RefSeq
Sequence: MGKPTSADLKLGLATGPVLFACQQFPEMNAMIMRRFSLPGDVDRARQYVLQSDGVQQTTYLAQQYCHEAIREISKLRPSPERDALIQLSEIVLTSpecifications
| PDSS1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant PDSS1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| COQ1/DPS/MGC70953/RP13-16H11.3/SPS/TPRT/TPT/hDPS1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 23590 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| PDSS1 | |
| PDSS1 | |
| Recombinant protein corresponding to amino acids of human PDSS1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only